Learn More
Medchemexpress LLC HY-10622 10mg Medchemexpress, PF-00446687 CAS:862281-92-3 Purity:>98%
SDP
Catalog No. 501968328
Encompass_Preferred
Medchemexpress, HY-10622 10mg PF-00446687 CAS:862281-92-3 PF-00446687 is a potent, selective melanocortin-4 receptor (MC4R) agonist with EC50 of 12 ± 1 nM. Pf-446687 is brain penetrant. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8328 Supplier Medchemexpress LLC Supplier No. HY1062210MG
Medchemexpress, HY-10622 10mg PF-00446687 CAS:862281-92-3 PF-00446687 is a potent, selective melanocortin-4 receptor (MC4R) agonist with EC50 of 12 ± 1 nM. Pf-446687 is brain penetrant. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.