Learn More
Medchemexpress LLC HY-10622 5mg Medchemexpress, PF-00446687 CAS:862281-92-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY106225MG
Encompass_Preferred
Medchemexpress, HY-10622 5mg PF-00446687 CAS:862281-92-3 PF-00446687 is a potent, selective melanocortin-4 receptor ( MC4R ) agonist with EC50 of 12 ± 1 nM [1] . Pf-446687 is brain penetrant [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2268