Learn More
Medchemexpress LLC PF-00446687 free bas 100mg | 862281-92-3 | 100MG
SDP
Catalog No. 5000244192
Encompass_Preferred
PF-00446687 is a potent selective melanocortin-4 receptor (MC4R) agonist with EC50 of 12 1 nM[1 Pf-446687 is brain penetrant[2
Catalog No. 50-002-44192 Supplier Medchemexpress LLC Supplier No. HY10622100MG
PF-00446687 is a potent selective melanocortin-4 receptor (MC4R) agonist with EC50 of 12 1 nM[1 Pf-446687 is brain penetrant[2