Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PF-00446687 free base | 862281-92-3 | 98.0% | 470.59 g/mol | C28H36F2N2O2 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY1062250MG

Encompass_Preferred

PF-00446687 free base is a non-peptide, brain-penetrant small-molecule agonist of the melanocortin-4 receptor (MC4R) used in research on melanocortin signaling and central nervous system pharmacology. It is supplied as a powder for in vitro and in vivo studies and demonstrates low-nanomolar potency and selectivity.

  • Selective melanocortin-4 receptor agonist with EC50 12 ± 1 nM.
  • Binding affinity (Ki) approximately 27 ± 4 nM.
  • Brain-penetrant, suitable for central nervous system studies.
  • Provided as a powder with solubility in DMSO 62.5 mg/mL (ultrasonic recommended).
  • Molecular formula C28H36F2N2O2 and molecular weight 470.59 g/mol.
  • High purity by HPLC (≈98.0% per COA).
  • Storage: powder -20°C (long-term) or 4°C (short-term).

Catalog No. 50-002-41861


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.