Learn More
Selleck Chemical LLC PF-00562271 Besylate-5mg
SDP
Catalog No. 502390004
Encompass_Preferred
PF-00562271 Besylate (PF-562271) is the benzenesulfonate salt of PF-562271, which is a potent, ATP-competitive, reversible inhibitor of FAK with IC50 of 1.5 nM, ~10-fold less potent for Pyk2 than FAK and >100-fold selectivity against other protein kinases, except for some CDKs. Phase 1.
Catalog No. 50-239-0004 Supplier Selleck Chemical LLC Supplier No. S26725MG
PF-00562271 Besylate (PF-562271) is the benzenesulfonate salt of PF-562271, which is a potent, ATP-competitive, reversible inhibitor of FAK with IC50 of 1.5 nM, ~10-fold less potent for Pyk2 than FAK and >100-fold selectivity against other protein kinases, except for some CDKs. Phase 1.