Learn More
Medchemexpress LLC Pf-02413873 | 936345-35-6 | 99.9% | 359.44 | 10 MG
SDP
Catalog No. 5000343927
Encompass_Preferred
PF-02413873 is a potent, selective, fully competitive, and orally active nonsteroidal progesterone receptor (PR) antagonist. It blocks progesterone binding and PR nuclear translocation, and inhibits endometrial growth in vivo.
  • Potent, selective, fully competitive, and orally active
  • Nonsteroidal progesterone receptor antagonist
  • Blocks progesterone binding and PR nuclear translocation
  • Inhibits endometrial growth in vivo
  • Ki: 2.6 nM (progesterone receptor)
Catalog No. 50-003-43927 Supplier Medchemexpress LLC Supplier No. HY1102810MG
PF-02413873 is a potent, selective, fully competitive, and orally active nonsteroidal progesterone receptor (PR) antagonist. It blocks progesterone binding and PR nuclear translocation, and inhibits endometrial growth in vivo.
  • Potent, selective, fully competitive, and orally active
  • Nonsteroidal progesterone receptor antagonist
  • Blocks progesterone binding and PR nuclear translocation
  • Inhibits endometrial growth in vivo
  • Ki: 2.6 nM (progesterone receptor)