Learn More
Medchemexpress LLC HY-11044 1mg Medchemexpress, PF-03654746 (Tosylate) CAS:1039399-17-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY110441MG
Encompass_Preferred
Medchemexpress, HY-11044 1mg PF-03654746 (Tosylate) CAS:1039399-17-1 PF-03654746 Tosylate is a potent and selective histamine H3 receptor antagonist with high brain penetration. PF-03654746 Tosylate reduces allergen-induced nasal symptoms. PF-03654746 Tosylate has potential for treatment of human cognitive disorders, improves cognitive efficacy and disease-modifying effects in Alzheimer's disease (AD). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8799