Learn More
Cayman Chemical PF-03814735, Purity: ≥95%, CAS: 942487-16-3, 50 mg
SDP
Catalog No. 502972229
Encompass_Preferred
A reversible inhibitor of both Aurora A and B kinases (IC50s = 0.8 and 5 nM, respectively); also inhibits FLT1, FAK, TrkA, Met, and FGFR1 (IC50s = 10, 22, 30, 100, and 100 nM, respectively)
Catalog No. 50-297-2229 Supplier Cayman Chemical Supplier No. 1501550 MG
A reversible inhibitor of both Aurora A and B kinases (IC50s = 0.8 and 5 nM, respectively); also inhibits FLT1, FAK, TrkA, Met, and FGFR1 (IC50s = 10, 22, 30, 100, and 100 nM, respectively)