Learn More
Apexbio Technology LLC PF-03814735, 50mg. Cas: 942487-16-3 MFCD: MFCD16659065. Aurora A/B inhibitor
SDP
Supplier:  Apexbio Technology LLC A412850
Encompass_Preferred
PF-03814735 is a potent, orally bioavailable, reversible inhibitor of both Aurora1 and Aurora2 kinases with IC50 values of 0.8nM and 5nM, respectively [1].PF-03814735 is an ATP competitive inhibitor of Aurora kinases. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4888