Learn More
Apexbio Technology LLC PF-04217903(Synonyms: PF04217903, PF 04217903, PF-4217903, PF4217903, MET inhibitor PF-04217903), 10mM (in 1mL DMSO), CAS: 956905-27-4.
SDP
Catalog No. 5000060540
Encompass_Preferred
PF-04217903 (CAS 956905-27-4) is a selective ATP-competitive inhibitor targeting the receptor tyrosine kinase c-Met with a reported Ki value of 4 8 nM Aberrant activation or amplification of c-Met signaling contributes significantly to tumor cell proliferation angiogenesis and metastasis PF-04217903 exhibits high selectivity for c-Met over numerous other kinases and inhibits various c-Met mutants including R988C V1092I H1094R and T1010I In cellular studies using lines such as GTL-16 and H1993 PF-04217903 inhibits proliferation and induces apoptosis In vivo it effectively reduces c-Met phosphorylation and tumor growth suggesting potential as a research tool for investigating c-Met-related oncogenic pathways
Catalog No. 50-000-60540 Supplier Apexbio Technology LLC Supplier No. A50965.1
PF-04217903 (CAS 956905-27-4) is a selective ATP-competitive inhibitor targeting the receptor tyrosine kinase c-Met with a reported Ki value of 4 8 nM Aberrant activation or amplification of c-Met signaling contributes significantly to tumor cell proliferation angiogenesis and metastasis PF-04217903 exhibits high selectivity for c-Met over numerous other kinases and inhibits various c-Met mutants including R988C V1092I H1094R and T1010I In cellular studies using lines such as GTL-16 and H1993 PF-04217903 inhibits proliferation and induces apoptosis In vivo it effectively reduces c-Met phosphorylation and tumor growth suggesting potential as a research tool for investigating c-Met-related oncogenic pathways