Learn More
Selleck Chemical LLC PF-04217903 50mg 956905-27-4
SDP
Catalog No. 508789
Encompass_Preferred
PF-04217903 is a selective ATP-competitive c-Met inhibitor with IC50 of 4.8 nM in A549 cell line, susceptible to oncogenic mutations (no activity to Y1230C mutant). Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-878-9 Supplier Selleck Chemical LLC Supplier No. S109450MG
PF-04217903 is a selective ATP-competitive c-Met inhibitor with IC50 of 4.8 nM in A549 cell line, susceptible to oncogenic mutations (no activity to Y1230C mutant). Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.