Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PF-04217903 mesylate | 956906-93-7 | 99.8% | 1 ML
SDP

Catalog No. 5000233919
Encompass_Preferred

PF-04217903 mesylate is the methanesulfonate salt of a potent ATP-competitive c-Met (HGFR) kinase inhibitor supplied as a high-purity research compound for biochemical and cell-based studies.

  • Potent ATP-competitive c-Met kinase inhibitor (Ki = 4.8 nM).
  • High purity, 99.82%.
  • Molecular weight 468.49 and formula C20H20N8O4S.
  • Available as 10 mM solution in DMSO, 1 mL, and as solids in multiple masses.
  • Suitable for in vitro biochemical and cellular assays.

Catalog No. 50-002-33919 Supplier Medchemexpress LLC Supplier No. HY12017A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

PF-04217903 mesylate is the methanesulfonate salt of a potent ATP-competitive c-Met (HGFR) kinase inhibitor supplied as a high-purity research compound for biochemical and cell-based studies.

  • Potent ATP-competitive c-Met kinase inhibitor (Ki = 4.8 nM).
  • High purity, 99.82%.
  • Molecular weight 468.49 and formula C20H20N8O4S.
  • Available as 10 mM solution in DMSO, 1 mL, and as solids in multiple masses.
  • Suitable for in vitro biochemical and cellular assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.