Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PF-04217903 mesylate | 956906-93-7 | 99.8% | 25 MG
SDP

Catalog No. 5000231859
Encompass_Preferred

PF-04217903 mesylate is the mesylate salt of PF-04217903, a potent ATP-competitive inhibitor of the c-Met (HGFR) tyrosine kinase intended for research use in biochemical and cell-based studies targeting MET signaling.

  • Potent ATP-competitive c-Met kinase inhibitor with reported Ki ≈ 4.8 nM.
  • High selectivity for c-Met, suitable for targeted biochemical and cell assays.
  • Supplied as a mesylate salt for improved stability in handling.
  • Small milligram pack sizes support preclinical and in vitro studies.
  • Molecular weight 468.49 g/mol; formula C20H20N8O4S.

Catalog No. 50-002-31859 Supplier Medchemexpress LLC Supplier No. HY12017A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PF-04217903 mesylate is the mesylate salt of PF-04217903, a potent ATP-competitive inhibitor of the c-Met (HGFR) tyrosine kinase intended for research use in biochemical and cell-based studies targeting MET signaling.

  • Potent ATP-competitive c-Met kinase inhibitor with reported Ki ≈ 4.8 nM.
  • High selectivity for c-Met, suitable for targeted biochemical and cell assays.
  • Supplied as a mesylate salt for improved stability in handling.
  • Small milligram pack sizes support preclinical and in vitro studies.
  • Molecular weight 468.49 g/mol; formula C20H20N8O4S.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.