Learn More
Cayman Chemical PF-04418948, Purity: ≥98%, CAS: 1078166-57-0, 100 mg
SDP
Catalog No. 502972251
Encompass_Preferred
An orally active, potent, and selective EP2 receptor antagonist (IC50 = 16 nM); over 1,000-fold less active at other prostanoid receptors, including other EP receptors
Catalog No. 50-297-2251 Supplier Cayman Chemical Supplier No. 15016100 MG
An orally active, potent, and selective EP2 receptor antagonist (IC50 = 16 nM); over 1,000-fold less active at other prostanoid receptors, including other EP receptors