Learn More
Apexbio Technology LLC PF 04418948, 1078166-57-0, MFCD24387541, 10 mg
SDP
Supplier:  Apexbio Technology LLC B572610
Encompass_Preferred
MF: C23H20FNO5, MW: 409.41, Solubility: insoluble in EtOH, Limited solubility in DMSO. PF-04418948 is a selective prostaglandin EP2 receptor antagonist with an IC50 of 16 nM, which has oral activity.
Catalog No. 50-115-2159