Learn More
Medchemexpress LLC HY-18966 5mg Medchemexpress, PF-04418948 CAS:1078166-57-0 Purity:>98%
SDP
Catalog No. 502030839
Encompass_Preferred
Medchemexpress, HY-18966 5mg PF-04418948 CAS:1078166-57-0 PF-04418948 is an orally active, potent and selective prostaglandin EP2 receptor antagonist with an IC50 of 16 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0839 Supplier Medchemexpress LLC Supplier No. HY189665MG
Medchemexpress, HY-18966 5mg PF-04418948 CAS:1078166-57-0 PF-04418948 is an orally active, potent and selective prostaglandin EP2 receptor antagonist with an IC50 of 16 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.