Learn More
Medchemexpress LLC HY-14376 50mg , PF-04457845 CAS:1020315-31-4 Purity:98%
SDP
Catalog No. 501493774
Encompass_Preferred
Medchemexpress, HY-14376 50mg PF-04457845 CAS:1020315-31-4 Formula:C23H20F3N5O2 IC50: 7.2±0.63 nM (hFAAH), 7.4±0.62 nM (rFAAH) Purity:98% PF-04457845 is a highly efficacious and selective FAAH inhibitor with IC 50 values is 7.2±0.63 nM and 7.4±0.62 nM for hFAAH and rFAAH , respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3774 Supplier Medchemexpress LLC Supplier No. HY1437650MG
Medchemexpress, HY-14376 50mg PF-04457845 CAS:1020315-31-4 Formula:C23H20F3N5O2 IC50: 7.2±0.63 nM (hFAAH), 7.4±0.62 nM (rFAAH) Purity:98% PF-04457845 is a highly efficacious and selective FAAH inhibitor with IC 50 values is 7.2±0.63 nM and 7.4±0.62 nM for hFAAH and rFAAH , respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.