Learn More
Cayman Chemical PF-04457845 50mg
SDP
Catalog No. 502985363
Encompass_Preferred
A selective, irreversible inhibitor of FAAH (IC50 = 7.2 nM); 0.1 mg/kg reduces inflammatory pain in a rat model
Catalog No. 50-298-5363 Supplier Cayman Chemical Supplier No. 1930650 MG
A selective, irreversible inhibitor of FAAH (IC50 = 7.2 nM); 0.1 mg/kg reduces inflammatory pain in a rat model