Learn More
Medchemexpress LLC HY-105283 10mg Medchemexpress, PF 04531083 CAS:1079400-07-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10528310MG
Encompass_Preferred
Medchemexpress, HY-105283 10mg PF 04531083 CAS:1079400-07-9 PF 04531083 is a selective NaV1.8 blocker, and used for the research of neuropathic/inflammatory pain. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8264