Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PF-04634817 succinate | 2140301-98-8 | 99.5% | 629.67 | C29H42F3N5O7 | 1 MG
SDP

Catalog No. 5000228159
Encompass_Preferred

PF-04634817 succinate is a research-grade small molecule antagonist of CCR2 and CCR5, provided for in vitro and in vivo pharmacology studies related to inflammation and metabolic disease models.

  • Dual CCR2/CCR5 antagonist with rat CCR2 IC50 20.8 nM and CCR5 IC50 470 nM.
  • High stated purity of 99.5% suitable for biological testing.
  • White to off-white solid with molecular weight 629.67 and formula C29H42F3N5O7.
  • Supplied in small milligram quantities for laboratory research.

Catalog No. 50-002-28159 Supplier Medchemexpress LLC Supplier No. HY117621A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PF-04634817 succinate is a research-grade small molecule antagonist of CCR2 and CCR5, provided for in vitro and in vivo pharmacology studies related to inflammation and metabolic disease models.

  • Dual CCR2/CCR5 antagonist with rat CCR2 IC50 20.8 nM and CCR5 IC50 470 nM.
  • High stated purity of 99.5% suitable for biological testing.
  • White to off-white solid with molecular weight 629.67 and formula C29H42F3N5O7.
  • Supplied in small milligram quantities for laboratory research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.