Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PF-04634817 succinate | 2140301-98-8 | 99.5% | 629.67 | C29H42F3N5O7 | 5 MG
SDP

Catalog No. 5000226729
Encompass_Preferred

PF-04634817 succinate is a research-grade small molecule dual CCR2/CCR5 chemokine receptor antagonist supplied as the succinate salt in a white to off-white solid. It is orally active in preclinical models, displays potent CCR2 activity (rat IC50 ≈ 20.8 nM) with lower CCR5 potency (rat IC50 ≈ 470 nM), and is intended for laboratory research use only.

  • Dual CCR2/CCR5 antagonist activity (rat CCR2 IC50 ≈ 20.8 nM; CCR5 IC50 ≈ 470 nM).
  • High purity (~99.5%).
  • Supplied as a succinate salt in solid form.
  • Molecular weight 629.67 Da.
  • Intended for in vitro and in vivo research; not for human use.
  • Available in 5 mg package size.

Catalog No. 50-002-26729 Supplier Medchemexpress LLC Supplier No. HY117621A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PF-04634817 succinate is a research-grade small molecule dual CCR2/CCR5 chemokine receptor antagonist supplied as the succinate salt in a white to off-white solid. It is orally active in preclinical models, displays potent CCR2 activity (rat IC50 ≈ 20.8 nM) with lower CCR5 potency (rat IC50 ≈ 470 nM), and is intended for laboratory research use only.

  • Dual CCR2/CCR5 antagonist activity (rat CCR2 IC50 ≈ 20.8 nM; CCR5 IC50 ≈ 470 nM).
  • High purity (~99.5%).
  • Supplied as a succinate salt in solid form.
  • Molecular weight 629.67 Da.
  • Intended for in vitro and in vivo research; not for human use.
  • Available in 5 mg package size.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.