Learn More
Selleck Chemical LLC PF-04691502 5mg 1013101-36-4 PF4691502
SDP
Supplier:  Selleck Chemical LLC S27435MG
Encompass_Preferred
PF-04691502 (PF4691502) is an ATP-competitive PI3K(?/?/?/?)/mTOR dual inhibitor with Ki of 1.8 nM/2.1 nM/1.6 nM/1.9 nM and 16 nM in cell-free assays, little activity against either Vps34, AKT, PDK1, p70S6K, MEK, ERK, p38, or JNK. PF-04691502 induces apoptosis. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-7971