Learn More
Cayman Chemical PF-04708671, Purity: ≥98%, CAS: 1255517-76-0, 100 mg
SDP
Catalog No. 502971599
Encompass_Preferred
A specific, cell-permeable inhibitor of S6K1 (IC50 = 160 nM); does not inhibit S6K2, MSK, or RSK, or many other unrelated kinases, under conditions in which it inhibits S6K1 activity
Catalog No. 50-297-1599 Supplier Cayman Chemical Supplier No. 15018100 MG
A specific, cell-permeable inhibitor of S6K1 (IC50 = 160 nM); does not inhibit S6K2, MSK, or RSK, or many other unrelated kinases, under conditions in which it inhibits S6K1 activity