Learn More
Cayman Chemical PF-04708671 50mg
SDP
Catalog No. 502972260
Encompass_Preferred
A specific, cell-permeable inhibitor of S6K1 (IC50 = 160 nM); does not inhibit S6K2, MSK, or RSK, or many other unrelated kinases, under conditions in which it inhibits S6K1 activity
Catalog No. 50-297-2260 Supplier Cayman Chemical Supplier No. 1501850 MG
A specific, cell-permeable inhibitor of S6K1 (IC50 = 160 nM); does not inhibit S6K2, MSK, or RSK, or many other unrelated kinases, under conditions in which it inhibits S6K1 activity