Learn More
Abcam PF-04929113 (SN x -5422), 100MG
Catalog No. 502682922
Encompass_Preferred
MW 521.5 Da, Purity >99%. Potent and selective Hsp90 inhibitor (IC₅₀ = 50 nM). A Her-2 degradation inducer (IC₅₀ = 37 nM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-2922 Supplier Abcam Supplier No. AB273614100MG
MW 521.5 Da, Purity >99%. Potent and selective Hsp90 inhibitor (IC₅₀ = 50 nM). A Her-2 degradation inducer (IC₅₀ = 37 nM).
The product is subject to the following: Abcam Restricted Use Statement