Learn More
Selleck Chemical LLC PF-04929113 (SNX-5422) S2656-5mg
SDP
Catalog No. 502173094
Encompass_Preferred
PF-04929113 (SNX-5422) is a potent and selective HSP90 inhibitor with Kd of 41 nM and induces Her-2 degradation with IC50 of 37 nM Phase 1/2
Catalog No. 50-217-3094 Supplier Selleck Chemical LLC Supplier No. S26565MG
PF-04929113 (SNX-5422) is a potent and selective HSP90 inhibitor with Kd of 41 nM and induces Her-2 degradation with IC50 of 37 nM Phase 1/2