Learn More
Medchemexpress LLC HY-108328 5mg Medchemexpress, PF-04937319 CAS:1245603-92-2 Purity:>98%
SDP
Catalog No. 501968529
Encompass_Preferred
Medchemexpress, HY-108328 5mg PF-04937319 CAS:1245603-92-2 PF-04937319 is a glucokinase activator (GKA) with EC50 value of 154.4  μM, one of the most promising strategies for the treatment of type 2 diabetes mellitus.PF-04937319 is designed to maintain glucose-lowering efficacy while mitigating the risk of hypoglycaemia observed with many other GKAs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8529 Supplier Medchemexpress LLC Supplier No. HY1083285MG
Medchemexpress, HY-108328 5mg PF-04937319 CAS:1245603-92-2 PF-04937319 is a glucokinase activator (GKA) with EC50 value of 154.4  μM, one of the most promising strategies for the treatment of type 2 diabetes mellitus.PF-04937319 is designed to maintain glucose-lowering efficacy while mitigating the risk of hypoglycaemia observed with many other GKAs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.