Learn More
Cayman Chemical PF-04937319 (hydrate), Purity: ≥98%, 10 mg
SDP
Catalog No. 502990484
Encompass_Preferred
A glucokinase activator (EC50s = 174, 407, and 269 nM for the recombinant human, rat, and dog enzymes, respectively); increases glucose-induced insulin secretion in isolated rat islets at 4 and 12 µM; reduces postprandial plasma glucose levels in rats from 3-100 mg/kg
Catalog No. 50-299-0484 Supplier Cayman Chemical Supplier No. 3092610 MG
A glucokinase activator (EC50s = 174, 407, and 269 nM for the recombinant human, rat, and dog enzymes, respectively); increases glucose-induced insulin secretion in isolated rat islets at 4 and 12 µM; reduces postprandial plasma glucose levels in rats from 3-100 mg/kg