Learn More
Medchemexpress LLC HY-15426 1mg Medchemexpress, PF-04957325 CAS:1305115-80-3 Purity:>98%
SDP
Catalog No. 502029122
Encompass_Preferred
Medchemexpress, HY-15426 1mg PF-04957325 CAS:1305115-80-3 PF-04957325 is a highly potent and selective PDE8 inhibitor, with IC50s of 0.7 nM and 0.3 nM for PDE8A and PDE8B, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9122 Supplier Medchemexpress LLC Supplier No. HY154261MG
Medchemexpress, HY-15426 1mg PF-04957325 CAS:1305115-80-3 PF-04957325 is a highly potent and selective PDE8 inhibitor, with IC50s of 0.7 nM and 0.3 nM for PDE8A and PDE8B, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.