Learn More
Medchemexpress LLC HY-100398 5mg Medchemexpress, PF-04979064 CAS:1220699-06-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1003985MG
Encompass_Preferred
Medchemexpress, HY-100398 5mg PF-04979064 CAS:1220699-06-8 PF-04979064 is a potent and selective PI3K / mTOR dual kinase inhibitor with K i s of 0.13 nM and 1.42 nM for PI3Kα and mTOR, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1809