Learn More
Cayman Chemical PF-04981517, Purity: ≥98%, CAS: 1390637-82-7, 5 mg
SDP
Catalog No. 502972007
Encompass_Preferred
A potent, selective, and time-dependent inhibitor of CYP3A4 (IC50 = 30 nM)
Catalog No. 50-297-2007 Supplier Cayman Chemical Supplier No. 150195 MG
A potent, selective, and time-dependent inhibitor of CYP3A4 (IC50 = 30 nM)