Learn More
Cayman Chemical PF-04995274, Purity: ≥98%, CAS: 1331782-27-4, 10 mg
SDP
Catalog No. 502940384
Encompass_Preferred
A partial agonist of the 5-HT4A, 5-HT4B, 5-HT4D, and 5-HT4E receptors (Kis = 0.36, 0.46, 0.15, and 0.32 nM, respectively, in a radioligand binding assay); increases cAMP levels in HEK293 cells expressing 5-HT4A, 5-HT4B, 5-HT4D, and 5-HT4E receptors (EC50s = 0.47, 0.36, 0.37, and 0.26 nM, respectively, for the human recombinant receptors); decreases scopolamine-induced increases in the distance traveled in the Morris water maze in rats at 0.032 mg/kg
Catalog No. 50-294-0384 Supplier Cayman Chemical Supplier No. 2877610 MG
A partial agonist of the 5-HT4A, 5-HT4B, 5-HT4D, and 5-HT4E receptors (Kis = 0.36, 0.46, 0.15, and 0.32 nM, respectively, in a radioligand binding assay); increases cAMP levels in HEK293 cells expressing 5-HT4A, 5-HT4B, 5-HT4D, and 5-HT4E receptors (EC50s = 0.47, 0.36, 0.37, and 0.26 nM, respectively, for the human recombinant receptors); decreases scopolamine-induced increases in the distance traveled in the Morris water maze in rats at 0.032 mg/kg