Learn More
Medchemexpress LLC HY-12942 10mg , PF-05175157 CAS:1301214-47-0 Purity:>98%
SDP
Catalog No. 502260683
Encompass_Preferred
HY-12942 10mg PF-05175157 CAS: 1301214-47-0 PF-05175157 is broad spectrum acetyl-CoA carboxylase (ACC) inhibitor with IC50s of 27.0, 33.0, 23.5 and 50.4 nM for ACC1 (human), ACC2 (human), ACC1 (rat), ACC2 (rat), respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-0683 Supplier Medchemexpress LLC Supplier No. HY1294210MG
HY-12942 10mg PF-05175157 CAS: 1301214-47-0 PF-05175157 is broad spectrum acetyl-CoA carboxylase (ACC) inhibitor with IC50s of 27.0, 33.0, 23.5 and 50.4 nM for ACC1 (human), ACC2 (human), ACC1 (rat), ACC2 (rat), respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.