Learn More
Cayman Chemical PF-05175157, Purity: ≥98%, CAS: 1301214-47-0, 5 mg
SDP
Catalog No. 502985853
Encompass_Preferred
An inhibitor of ACC1 and 2 (IC50s = 27, 33, 23.5, and 50.4 nM for human ACC1, human ACC2, rat ACC1, and rat ACC2, respectively); inhibits formation of malonyl-CoA in primary rat hepatocytes (EC50 = 29.9 nM); inhibits formation of malonyl-CoA in the liver and skeletal muscle, inhibits hepatic de novo lipogenesis, and lowers the respiratory exchange rate in rats (ED50s = 6, 11, 7.8, and 3.5 mg/kg, respectively)
Catalog No. 50-298-5853 Supplier Cayman Chemical Supplier No. 217785 MG
An inhibitor of ACC1 and 2 (IC50s = 27, 33, 23.5, and 50.4 nM for human ACC1, human ACC2, rat ACC1, and rat ACC2, respectively); inhibits formation of malonyl-CoA in primary rat hepatocytes (EC50 = 29.9 nM); inhibits formation of malonyl-CoA in the liver and skeletal muscle, inhibits hepatic de novo lipogenesis, and lowers the respiratory exchange rate in rats (ED50s = 6, 11, 7.8, and 3.5 mg/kg, respectively)