Learn More
Medchemexpress LLC HY-111371 5mg Medchemexpress, PF-05180999 CAS:1394033-54-5 Purity:>98%
SDP
Catalog No. 501968843
Encompass_Preferred
Medchemexpress, HY-111371 5mg PF-05180999 CAS:1394033-54-5 PF-05180999 is a phosphodiesterase 2A (PDE2A) inhibitor, with an IC50 of 1.6 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8843 Supplier Medchemexpress LLC Supplier No. HY1113715MG
Medchemexpress, HY-111371 5mg PF-05180999 CAS:1394033-54-5 PF-05180999 is a phosphodiesterase 2A (PDE2A) inhibitor, with an IC50 of 1.6 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.