Learn More
Medchemexpress LLC HY-19562 10mg , PF-06260933 CAS:1811510-56-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1956210MG
Encompass_Preferred
HY-19562 10mg PF-06260933 CAS: 1811510-56-1 PF-06260933 is an orally active and highly selective inhibitor of MAP4K4 with IC50s of 3.7 and 160 nM for kinase and cell, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1042