Learn More
Selleck Chemical LLC PF-06273340 10mg 1402438-74-7
SDP
Supplier:  Selleck Chemical LLC S840710MG
Encompass_Preferred
PF-06273340 is a highly potent, kinases elective, well-tolerated pan-Trk inhibitor with IC50 values of 6, 4, 3 nM for TrkA, TrkB, Trk C, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8220