Learn More
Medchemexpress LLC HY-19321 5mg Medchemexpress, PF-06282999 CAS:1435467-37-0 Purity:>98%
SDP
Catalog No. 502082117
Encompass_Preferred
Medchemexpress, HY-19321 5mg PF-06282999 CAS:1435467-37-0 PF-06282999 is a potent and selective myeloperoxidase inhibitor which is potential useful for the treatment of cardiovascular diseases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2117 Supplier Medchemexpress LLC Supplier No. HY193215MG
Medchemexpress, HY-19321 5mg PF-06282999 CAS:1435467-37-0 PF-06282999 is a potent and selective myeloperoxidase inhibitor which is potential useful for the treatment of cardiovascular diseases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.