Learn More
Medchemexpress LLC HY-19947 5mg Medchemexpress, PF-06291874 CAS:1393124-08-7 Purity:>98%
SDP
Catalog No. 502082200
Encompass_Preferred
Medchemexpress, HY-19947 5mg PF-06291874 CAS:1393124-08-7 PF-06291874 is a highly potent, non-peptide and orally active glucagon receptor antagonist. PF-06291874 is under the study for type 2 diabetes mellitus (T2DM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2200 Supplier Medchemexpress LLC Supplier No. HY199475MG
Medchemexpress, HY-19947 5mg PF-06291874 CAS:1393124-08-7 PF-06291874 is a highly potent, non-peptide and orally active glucagon receptor antagonist. PF-06291874 is under the study for type 2 diabetes mellitus (T2DM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.