Learn More
TARGETMOL CHEMICALS INC PF-06305591 10MG
SDP
Catalog No. 502581454
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PF-06305591 is an effective and selective blocker of voltage-gated sodium channel NaV1.8 (IC50 = 15 nM). purity: 99%
Catalog No. 50-258-1454 Supplier TARGETMOL CHEMICALS INC Supplier No. T1242410MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PF-06305591 is an effective and selective blocker of voltage-gated sodium channel NaV1.8 (IC50 = 15 nM). purity: 99%