Learn More
Medchemexpress LLC PF-06305591 dihydrat | 5MG
SDP
Catalog No. 5000307514
Encompass_Preferred
PF-06305591 dihydrate is a potent and highly selective voltage gated sodium channel NaV1 8 blocker with an IC50 of 15 nM An excellent preclinical in vitro ADME and safety profile[1]
Catalog No. 50-003-07514 Supplier Medchemexpress LLC Supplier No. HY114301A5MG
PF-06305591 dihydrate is a potent and highly selective voltage gated sodium channel NaV1 8 blocker with an IC50 of 15 nM An excellent preclinical in vitro ADME and safety profile[1]