Learn More
Medchemexpress LLC PF-06380101 (Aur0101) | 1436391-86-4 | 100.0% | 743.01 g/mol | C39H62N6O6S | 25 MG
SDP
Catalog No. 5000203210
Encompass_Preferred
PF-06380101 (Aur0101) is an auristatin-class microtubule inhibitor and a cytotoxic dolastatin 10 analogue developed for use as an antibody-drug conjugate (ADC) payload and in oncology research. It shows potent activity in tumor cell proliferation assays and has differential ADME properties compared with other synthetic auristatin analogues. Supplied as a solid for research use only.
  • Potent microtubule inhibitor for cancer research.
  • Suitable as antibody-drug conjugate (ADC) payload.
  • High reported purity for analytical and experimental use.
  • White to off-white solid form, easy to handle.
  • Recommended storage: powder at -20°C or 4°C; in solvent at -80°C.
  • Manufacturer-provided datasheet includes CAS number, formula, and molecular weight.
Catalog No. 50-002-03210 Supplier Medchemexpress LLC Supplier No. HY1252225MG
PF-06380101 (Aur0101) is an auristatin-class microtubule inhibitor and a cytotoxic dolastatin 10 analogue developed for use as an antibody-drug conjugate (ADC) payload and in oncology research. It shows potent activity in tumor cell proliferation assays and has differential ADME properties compared with other synthetic auristatin analogues. Supplied as a solid for research use only.
  • Potent microtubule inhibitor for cancer research.
  • Suitable as antibody-drug conjugate (ADC) payload.
  • High reported purity for analytical and experimental use.
  • White to off-white solid form, easy to handle.
  • Recommended storage: powder at -20°C or 4°C; in solvent at -80°C.
  • Manufacturer-provided datasheet includes CAS number, formula, and molecular weight.