Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PF-06380101 (Aur0101) | 1436391-86-4 | MFCD28385873 | 100.0% | 743.01 g/mol | C39H62N6O6S | 100 MG
SDP

Supplier:  Medchemexpress LLC HY12522100MG

Encompass_Preferred

PF-06380101 (Aur0101) is an auristatin-class microtubule inhibitor and a dolastatin 10 analogue used as a cytotoxic payload in antibody-drug conjugates and for tumor cell proliferation assays. It is supplied as a solid powder for research use only and displays potent antimitotic activity with distinct ADME properties compared with related analogues.

  • Auristatin-class microtubule inhibitor with potent antimitotic activity.
  • High purity of 99.95%.
  • Molecular weight 743.01 g/mol and formula C39H62N6O6S.
  • Solid, white to off-white powder appearance.
  • Powder stability: -20°C for 3 years, 4°C for 2 years.
  • Intended for research use only and preparation of antibody-drug conjugates.

Catalog No. 50-002-02091


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.