Learn More
Medchemexpress LLC PF-06380101-d8 | 1436391-86-4 | 99.3% | 751.06 | 10 MG
SDP
Catalog No. 5000436547
Encompass_Preferred
PF-06380101-d8 is a deuterium labeled compound, an Auristatin microtubule inhibitor, and a cytotoxic Dolastatin 10 analogue.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-36547 Supplier Medchemexpress LLC Supplier No. HY12522S10MG
PF-06380101-d8 is a deuterium labeled compound, an Auristatin microtubule inhibitor, and a cytotoxic Dolastatin 10 analogue.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.