Learn More
Medchemexpress LLC PF-06380101-d8 (Aur0101-d8) | 00-00-0 | MFCD28385873 | 99.3% | 751.06 g·mol⁻¹ | C39H54D8N6O6S | 5 MG
SDP
Catalog No. 5000264665
Encompass_Preferred
PF-06380101-d8 (Aur0101-d8) is a deuterium-labeled form of an auristatin-class microtubule inhibitor supplied for research use. It serves as an isotopically labeled reference or tracer for analytical quantitation, ADC payload research, and ADME/PK studies. The material is provided as a high-purity solid with defined storage recommendations for stable handling and long-term conservation.
  • Deuterium-labeled isotopologue for tracer and quantitation studies.
  • High purity suitable for analytical standards and research use.
  • Specified storage guidance for stability in solution and solid form.
  • Applicable to antibody-drug conjugate payload research and ADME studies.
  • Small pack size suitable for scarce or specialized research needs.
Catalog No. 50-002-64665 Supplier Medchemexpress LLC Supplier No. HY12522S5MG
PF-06380101-d8 (Aur0101-d8) is a deuterium-labeled form of an auristatin-class microtubule inhibitor supplied for research use. It serves as an isotopically labeled reference or tracer for analytical quantitation, ADC payload research, and ADME/PK studies. The material is provided as a high-purity solid with defined storage recommendations for stable handling and long-term conservation.
  • Deuterium-labeled isotopologue for tracer and quantitation studies.
  • High purity suitable for analytical standards and research use.
  • Specified storage guidance for stability in solution and solid form.
  • Applicable to antibody-drug conjugate payload research and ADME studies.
  • Small pack size suitable for scarce or specialized research needs.