Learn More
Medchemexpress LLC HY-103683 10mg , PF-06409577 CAS:1467057-23-3 Purity:>98%
SDP
Catalog No. 502260451
Encompass_Preferred
HY-103683 10mg PF-06409577 CAS: 1467057-23-3 PF-06409577 is a potent and selective allosteric activator of AMPK α1β1γ1 isoform with an EC50 of 7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-0451 Supplier Medchemexpress LLC Supplier No. HY10368310MG
HY-103683 10mg PF-06409577 CAS: 1467057-23-3 PF-06409577 is a potent and selective allosteric activator of AMPK α1β1γ1 isoform with an EC50 of 7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.