Learn More
TARGETMOL CHEMICALS INC PF-06424439 METHANESULFO 25MG
SDP
Catalog No. 502581453
Encompass_Preferred
Also available in 1 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PF-06424439 methanesulfonate is an oral potent and selective imidazopyridine diacylglycerol acyltransferase 2 (DGAT2) inhibitor(IC50 of 14 nM)[1]. PF-06424439 methanesulfonate is slowly reversible time-dependent inhibitor which inhibits DGAT2 in a noncompetitive mode with respect to the acyl-CoA substrate[2]. purity: 99%
Catalog No. 50-258-1453 Supplier TARGETMOL CHEMICALS INC Supplier No. T1242525MG
Also available in 1 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PF-06424439 methanesulfonate is an oral potent and selective imidazopyridine diacylglycerol acyltransferase 2 (DGAT2) inhibitor(IC50 of 14 nM)[1]. PF-06424439 methanesulfonate is slowly reversible time-dependent inhibitor which inhibits DGAT2 in a noncompetitive mode with respect to the acyl-CoA substrate[2]. purity: 99%