Learn More
Medchemexpress LLC HY-108341A 5mg Medchemexpress, PF-06424439 (methanesulfonate) CAS:1469284-79-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY108341A5MG
Encompass_Preferred
Medchemexpress, HY-108341A 5mg PF-06424439 (methanesulfonate) CAS:1469284-79-4 PF-06424439 methanesulfonate is an oral, potent and selective imidazopyridine diacylglycerol acyltransferase 2 (DGAT2) inhibitor with an IC50 of 14 nM. PF-06424439 methanesulfonate is slowly reversible, time-dependent inhibitor, which inhibits DGAT2 in a noncompetitive mode with respect to the acyl-CoA substrate. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8535