Learn More
Medchemexpress LLC PF-06424439 methanesulfonate | 1469284-79-4 | >98.0% | 536.05 g/mol | C22H26ClN7O · CH3SO3H | 10 MG
SDP
Catalog No. 5000244064
Encompass_Preferred
PF-06424439 methanesulfonate is the mesylate salt of a potent, selective diacylglycerol acyltransferase 2 (DGAT2) inhibitor used in biochemical and preclinical research. It shows low-nanomolar in vitro potency and has been reported to reduce plasma and hepatic triglycerides in animal models. Supplied as a research-grade solid for laboratory assays.
  • Potent, selective DGAT2 inhibition (IC50 ≈ 14 nM).
  • Available as the methanesulfonate (mesylate) salt for improved handling.
  • Documented activity in preclinical models reducing plasma and hepatic triglycerides.
  • High purity suitable for research applications (>98% by HPLC).
  • Provided in small research pack sizes for assay development and pharmacology studies.
Catalog No. 50-002-44064 Supplier Medchemexpress LLC Supplier No. HY108341A10MG
PF-06424439 methanesulfonate is the mesylate salt of a potent, selective diacylglycerol acyltransferase 2 (DGAT2) inhibitor used in biochemical and preclinical research. It shows low-nanomolar in vitro potency and has been reported to reduce plasma and hepatic triglycerides in animal models. Supplied as a research-grade solid for laboratory assays.
  • Potent, selective DGAT2 inhibition (IC50 ≈ 14 nM).
  • Available as the methanesulfonate (mesylate) salt for improved handling.
  • Documented activity in preclinical models reducing plasma and hepatic triglycerides.
  • High purity suitable for research applications (>98% by HPLC).
  • Provided in small research pack sizes for assay development and pharmacology studies.