Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PF-06424439 methanesulfonate | 1469284-79-4 | 99.9% | 536.05 | C23H30ClN7O4S | 50 MG
SDP

Catalog No. 5000243402
Encompass_Preferred

PF-06424439 methanesulfonate is an orally active, potent, and selective imidazopyridine inhibitor of diacylglycerol acyltransferase 2 (DGAT2) used in preclinical research to study lipid metabolism and triglyceride synthesis. The methanesulfonate salt is supplied as a characterized solid with supporting analytical documentation.

  • Potent, selective DGAT2 inhibitor with IC50 of 14 nM.
  • High purity suitable for research applications (reported 99.85%).
  • Molecular weight 536.05, consistent with analytical characterization.
  • Solid appearance: white to yellow.
  • Storage recommendations provided for solid and solution states to preserve stability.

Catalog No. 50-002-43402 Supplier Medchemexpress LLC Supplier No. HY108341A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PF-06424439 methanesulfonate is an orally active, potent, and selective imidazopyridine inhibitor of diacylglycerol acyltransferase 2 (DGAT2) used in preclinical research to study lipid metabolism and triglyceride synthesis. The methanesulfonate salt is supplied as a characterized solid with supporting analytical documentation.

  • Potent, selective DGAT2 inhibitor with IC50 of 14 nM.
  • High purity suitable for research applications (reported 99.85%).
  • Molecular weight 536.05, consistent with analytical characterization.
  • Solid appearance: white to yellow.
  • Storage recommendations provided for solid and solution states to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.